Product Details

SNP ID
rs16835516
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:153734884 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATCTTTAAGGCCATCCCATTAGA[A/G]TAGAAGTAGTGTCTGGGAGTGGAGA
Phenotype
MIM: 611347
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INTS3 PubMed Links

Gene Details

Gene
INTS3
Gene Name
integrator complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324475.1 Intron NP_001311404.1
NM_023015.4 Intron NP_075391.3
XM_005245459.2 Intron XP_005245516.1
XM_005245461.2 Intron XP_005245518.1
XM_006711490.3 Intron XP_006711553.1
XM_011509906.2 Intron XP_011508208.1
XM_017002132.1 Intron XP_016857621.1

View Full Product Details