Product Details

SNP ID
rs16834457
Assay Type
Functionally Tested
NCBI dbSNP Submissions
24
Location
Chr.1:152759925 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTCTGTTCAAAGCCAGGCTCCA[C/T]GCCAATCTGAGGTGTCCTACGTGCA
Phenotype
MIM: 613260
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KPRP PubMed Links

Gene Details

Gene
KPRP
Gene Name
keratinocyte proline rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025231.1 436 Missense Mutation CGC,TGC R113C NP_001020402.1
XM_011509571.1 436 Missense Mutation CGC,TGC R113C XP_011507873.1

View Full Product Details