Product Details
- SNP ID
-
rs17600020
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:150699411 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCTTCTACTTCAGTTTTGATCTC[C/T]AGAAAGCTCAGATCAAATTTGTGGC
- Phenotype
-
MIM: 612208
MIM: 609824
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GOLPH3L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74124973] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GOLPH3L
- Gene Name
- golgi phosphoprotein 3 like
There are no transcripts associated with this gene.
- Gene
- HORMAD1
- Gene Name
- HORMA domain containing 1
View Full Product Details