Product Details

SNP ID
rs17477369
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:115033790 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATCAACGATGAAAGAGAGGAGGG[A/T]CTCACTTTTGTCTCTGTAGAATTCA
Phenotype
MIM: 188540
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TSHB PubMed Links

Gene Details

Gene
TSHB
Gene Name
thyroid stimulating hormone beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000549.4 Intron NP_000540.2
NM_001277991.1 Intron NP_001264920.1
XM_011542065.2 Intron XP_011540367.1

View Full Product Details