Product Details

SNP ID
rs16982282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26527376 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCACAGCGAGGGCCCAGCACTTCA[C/T]GGTTTCCAAGCACTCCCACCTGGGC
Phenotype
MIM: 603126
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPST2 PubMed Links

Gene Details

Gene
TPST2
Gene Name
tyrosylprotein sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008566.1 Intron NP_001008566.1
NM_003595.3 Intron NP_003586.3
XM_006724338.3 Intron XP_006724401.1

View Full Product Details