Product Details

SNP ID
rs34066050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38216597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTAGGGCTTCTGGTGGCGGGGA[A/G]GGCTAGTGCACATCACCAGCTATTG
Phenotype
MIM: 604877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFF PubMed Links

Gene Details

Gene
MAFF
Gene Name
MAF bZIP transcription factor F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161572.1 1288 Intron NP_001155044.1
NM_001161573.1 1288 Intron NP_001155045.1
NM_001161574.1 1288 Intron NP_001155046.1
NM_012323.3 1288 Intron NP_036455.1
Gene
SNORA92
Gene Name
small nucleolar RNA, H/ACA box 92
There are no transcripts associated with this gene.

Gene
TMEM184B
Gene Name
transmembrane protein 184B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195071.1 1288 Intron NP_001182000.1
NM_001195072.1 1288 Intron NP_001182001.1
NM_012264.4 1288 Intron NP_036396.2
XM_011530112.2 1288 Intron XP_011528414.2
XM_011530113.1 1288 Intron XP_011528415.1
XM_011530114.1 1288 Intron XP_011528416.1
XM_011530115.2 1288 Intron XP_011528417.1
XM_011530116.2 1288 UTR 3 XP_011528418.2
XM_011530117.1 1288 Intron XP_011528419.1
XM_011530118.1 1288 Intron XP_011528420.1
XM_011530119.1 1288 Intron XP_011528421.1
XM_017028754.1 1288 Intron XP_016884243.1
XM_017028755.1 1288 Intron XP_016884244.1
XM_017028756.1 1288 Intron XP_016884245.1
XM_017028757.1 1288 Intron XP_016884246.1
XM_017028758.1 1288 UTR 3 XP_016884247.1
XM_017028759.1 1288 Intron XP_016884248.1

View Full Product Details