Product Details

SNP ID
rs17056989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27311588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTGTTACTGGAAACCTACTTCCG[A/G]CTGCAAATGGGAAAAGGAGCCTCTA
Phenotype
MIM: 601212 MIM: 617007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTK2B PubMed Links

Gene Details

Gene
PTK2B
Gene Name
protein tyrosine kinase 2 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004103.4 107 Intron NP_004094.3
NM_173174.2 107 UTR 5 NP_775266.1
NM_173175.2 107 Intron NP_775267.1
NM_173176.2 107 Intron NP_775268.1
XM_005273447.4 107 Intron XP_005273504.1
XM_005273448.4 107 Intron XP_005273505.1
XM_011544441.2 107 Intron XP_011542743.1
XM_011544442.2 107 Intron XP_011542744.1
XM_017013214.1 107 Intron XP_016868703.1
XM_017013215.1 107 Intron XP_016868704.1
XM_017013216.1 107 Intron XP_016868705.1
Gene
TRIM35
Gene Name
tripartite motif containing 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304495.1 107 Intron NP_001291424.1
NM_171982.4 107 Intron NP_741983.2
XM_005273452.3 107 Intron XP_005273509.1
XM_011544453.1 107 Intron XP_011542755.1

View Full Product Details