Product Details

SNP ID
rs17754651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32598082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTAGTTCTCAGATGTGTTCAC[A/G]CTCAGCATCATCTTGTTGCATCCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD27 PubMed Links

Gene Details

Gene
ANKRD27
Gene Name
ankyrin repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032139.2 3372 UTR 3 NP_115515.2
XM_006723412.2 3372 UTR 3 XP_006723475.1
XM_017027349.1 3372 UTR 3 XP_016882838.1

View Full Product Details