Product Details

SNP ID
rs16919376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93788092 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACAGAGATGTGTGTTCTCTATGA[C/T]GTTCTCAGTATTGTTAGGGATAAAA
Phenotype
MIM: 603810
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED17 PubMed Links

Gene Details

Gene
MED17
Gene Name
mediator complex subunit 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004268.4 617 Silent Mutation GAC,GAT D114D NP_004259.3
XM_011543068.2 617 Silent Mutation GAC,GAT D114D XP_011541370.1

View Full Product Details