Product Details

SNP ID
rs17532330
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:103571101 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTAACCAGTTGTTTCTGTCCTGT[C/T]GGTCTGTCAGGGTTGGAAAGTCCAA
Phenotype
MIM: 104660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTG1P4 PubMed Links

Gene Details

Gene
ACTG1P4
Gene Name
actin gamma 1 pseudogene 4
There are no transcripts associated with this gene.

Gene
AMY2B
Gene Name
amylase, alpha 2B (pancreatic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020978.4 Intron NP_066188.1

View Full Product Details