Product Details

SNP ID
rs17365723
Assay Type
Functionally Tested
NCBI dbSNP Submissions
39
Location
Chr.1:84503336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTAATAGCATCACAAATGCTCTC[C/T]ATACCAAGAACCCCTGCAGATTACA
Phenotype
MIM: 600874
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GNG5 PubMed Links

Gene Details

Gene
GNG5
Gene Name
G protein subunit gamma 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005274.2 Intron NP_005265.1
Gene
RPF1
Gene Name
ribosome production factor 1 homolog
There are no transcripts associated with this gene.

Gene
SPATA1
Gene Name
spermatogenesis associated 1
There are no transcripts associated with this gene.

View Full Product Details