Product Details

SNP ID
rs17347170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:78350925 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGTTTTTGGAGACAGGCTCTTT[C/T]AGTGCATAGTCGTGGATGACACCTT
Phenotype
MIM: 606382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGI2 PubMed Links

Gene Details

Gene
MAGI2
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301128.1 Intron NP_001288057.1
NM_012301.3 Intron NP_036433.2
XM_011516718.1 Intron XP_011515020.1
XM_011516719.2 Intron XP_011515021.1
XM_011516720.2 Intron XP_011515022.1
XM_011516726.2 Intron XP_011515028.1
XM_011516728.1 Intron XP_011515030.1
XM_017012840.1 Intron XP_016868329.1
XM_017012841.1 Intron XP_016868330.1
XM_017012842.1 Intron XP_016868331.1
XM_017012843.1 Intron XP_016868332.1
XM_017012844.1 Intron XP_016868333.1
XM_017012845.1 Intron XP_016868334.1
XM_017012846.1 Intron XP_016868335.1
XM_017012847.1 Intron XP_016868336.1
XM_017012848.1 Intron XP_016868337.1
XM_017012849.1 Intron XP_016868338.1
XM_017012850.1 Intron XP_016868339.1
XM_017012851.1 Intron XP_016868340.1
XM_017012852.1 Intron XP_016868341.1
Gene
RPL13AP17
Gene Name
ribosomal protein L13a pseudogene 17
There are no transcripts associated with this gene.

View Full Product Details