Product Details
- SNP ID
-
rs5029981
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:186720525 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTGGGCCAAGCCAAAGAGCTGGC[A/G]AGGGGAGGGGGTGGGTGCTTCCATT
- Phenotype
-
MIM: 612358
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KNG1
PubMed Links
Gene Details
- Gene
- KNG1
- Gene Name
- kininogen 1
View Full Product Details