Product Details

SNP ID
rs17694337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52860551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAGATCAAAAGATTGAATTAC[A/G]CACCCCGTACCCCAAAATCATATTG
Phenotype
MIM: 607746
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FERMT2 PubMed Links

Gene Details

Gene
FERMT2
Gene Name
fermitin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134999.1 Intron NP_001128471.1
NM_001135000.1 Intron NP_001128472.1
NM_006832.2 Intron NP_006823.1
XM_005267285.2 Intron XP_005267342.1
XM_006720008.2 Intron XP_006720071.1
XM_006720009.2 Intron XP_006720072.1
XM_006720010.2 Intron XP_006720073.1

View Full Product Details