Product Details
- SNP ID
-
rs17128455
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:55414300 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTACTTGTAACATCTACAATTAAAC[A/G]TAGAAGAATCCAGCTGTTTCTGTGC
- Phenotype
-
MIM: 613515
MIM: 608964
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATG14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79919449] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATG14
- Gene Name
- autophagy related 14
There are no transcripts associated with this gene.
- Gene
- TBPL2
- Gene Name
- TATA-box binding protein like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199047.2 |
1207 |
UTR 3 |
|
|
NP_950248.1 |
View Full Product Details