Product Details

SNP ID
rs8008130
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55787267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTAGATGCATCACTCTGTTGAC[A/C]AAAAGAGTCAAACTCTGTAAAATGG
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
LINC00520 PubMed Links

Gene Details

Gene
LINC00520
Gene Name
long intergenic non-protein coding RNA 520
There are no transcripts associated with this gene.

View Full Product Details