Product Details

SNP ID
rs16956603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66512887 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGTTTGAGCCCAAACAGATATTC[C/T]TATGGCCACGCACAAAAGCTCATGC
Phenotype
MIM: 188250
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TK2 PubMed Links

Gene Details

Gene
TK2
Gene Name
thymidine kinase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172643.1 Intron NP_001166114.1
NM_001172644.1 Intron NP_001166115.1
NM_001172645.1 Intron NP_001166116.1
NM_001271934.1 Intron NP_001258863.1
NM_001271935.1 Intron NP_001258864.1
NM_001272050.1 Intron NP_001258979.1
NM_004614.4 Intron NP_004605.4

View Full Product Details