Product Details

SNP ID
rs16923022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23103744 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATAACCGACTAGCTCACAGTT[C/T]TTTTCTGCCCAATGCCTCAACTACT
Phenotype
MIM: 613782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSRB2 PubMed Links

Gene Details

Gene
MSRB2
Gene Name
methionine sulfoxide reductase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012228.3 Intron NP_036360.3
XM_011519426.2 Intron XP_011517728.1

View Full Product Details