Product Details

SNP ID
rs17361990
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:179101902 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAATGACAGTGAAGTTCAAGTG[A/G]ACTACAGTTGTGCTGAACAGAGCAA
Phenotype
MIM: 164690 MIM: 607555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABL2 PubMed Links

Gene Details

Gene
ABL2
Gene Name
ABL proto-oncogene 2, non-receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136000.2 8993 UTR 3 NP_001129472.1
NM_001136001.1 8993 Intron NP_001129473.1
NM_001168236.1 8993 UTR 3 NP_001161708.1
NM_001168237.1 8993 UTR 3 NP_001161709.1
NM_001168238.1 8993 UTR 3 NP_001161710.1
NM_001168239.1 8993 UTR 3 NP_001161711.1
NM_005158.4 8993 UTR 3 NP_005149.4
NM_007314.3 8993 UTR 3 NP_009298.1
XM_005245088.2 8993 Intron XP_005245145.1
XM_017001035.1 8993 Intron XP_016856524.1
Gene
TOR3A
Gene Name
torsin family 3 member A
There are no transcripts associated with this gene.

View Full Product Details