Product Details
- SNP ID
-
rs16852778
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
33
- Location
-
Chr.1:204082313 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGCTCTGGAGACCCTGGCCAGGCT[C/T]CTCCAACCCATGGGAACCAGGCAGG
- Phenotype
-
MIM: 604748
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SOX13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112497628] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SOX13
- Gene Name
- SRY-box 13
View Full Product Details