Product Details

SNP ID
rs6754353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:45388884 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAAAAAAATCTATAAAATGGAA[C/G]GATTAAACACTGCCCAGCCCTACTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SRBD1 PubMed Links

Gene Details

Gene
SRBD1
Gene Name
S1 RNA binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018079.4 3044 UTR 3 NP_060549.4
XM_011532946.1 3044 UTR 3 XP_011531248.1
XM_017004417.1 3044 Intron XP_016859906.1
XM_017004418.1 3044 UTR 3 XP_016859907.1

View Full Product Details