Product Details

SNP ID
rs16867024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9409549 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTTTCAGTACAACACTTAGACT[C/G]GTTGTAGCTTTTATGGTTAAGACCA
Phenotype
MIM: 603817 MIM: 607153
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASAP2 PubMed Links

Gene Details

Gene
ASAP2
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
There are no transcripts associated with this gene.

Gene
ITGB1BP1
Gene Name
integrin subunit beta 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319066.1 Intron NP_001305995.1
NM_001319067.1 Intron NP_001305996.1
NM_001319068.1 Intron NP_001305997.1
NM_001319069.1 Intron NP_001305998.1
NM_001319070.1 Intron NP_001305999.1
NM_004763.4 Intron NP_004754.1
NM_022334.4 Intron NP_071729.1
XM_005246183.4 Intron XP_005246240.1
XM_005246184.4 Intron XP_005246241.1
XM_005246185.4 Intron XP_005246242.1
XM_005246189.4 Intron XP_005246246.1
XM_006711903.3 Intron XP_006711966.1
XM_011510416.2 Intron XP_011508718.1
XM_017005267.1 Intron XP_016860756.1
XM_017005268.1 Intron XP_016860757.1
XM_017005269.1 Intron XP_016860758.1
XM_017005270.1 Intron XP_016860759.1

View Full Product Details