Product Details

SNP ID
rs17856349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46084357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGATTGTTCTGAGATCTTTCC[A/C]TCAGCCCTATTTGGAAATAAAAGCA
Phenotype
MIM: 164360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP5A1 PubMed Links

Gene Details

Gene
ATP5A1
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001935.2 1709 Missense Mutation GAG,GAT E479D NP_001001935.1
NM_001001937.1 1709 Missense Mutation GAG,GAT E529D NP_001001937.1
NM_001257334.1 1709 Missense Mutation GAG,GAT E507D NP_001244263.1
NM_001257335.1 1709 Missense Mutation GAG,GAT E479D NP_001244264.1
NM_004046.5 1709 Missense Mutation GAG,GAT E529D NP_004037.1
XM_017025789.1 1709 Missense Mutation AGG,ATG R576M XP_016881278.1

View Full Product Details