Product Details

SNP ID
rs17363069
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:29193512 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCCTCAGGGCCCTGATGACCTCA[C/G]CCTGGCATTCACTCTTCCATGCAGC
Phenotype
MIM: 608205
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MECR PubMed Links

Gene Details

Gene
MECR
Gene Name
mitochondrial trans-2-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024732.2 1684 UTR 3 NP_001019903.2
NM_016011.3 1684 UTR 3 NP_057095.3
XM_005245885.2 1684 UTR 3 XP_005245942.1
XM_005245887.2 1684 Intron XP_005245944.1
XM_011541539.1 1684 Intron XP_011539841.1
XM_011541540.1 1684 Intron XP_011539842.1
XM_011541541.1 1684 Intron XP_011539843.1
XM_011541543.1 1684 Intron XP_011539845.1
XM_011541545.1 1684 Intron XP_011539847.1
XM_011541546.1 1684 Intron XP_011539848.1
XM_011541547.1 1684 Intron XP_011539849.1
XM_011541549.1 1684 Intron XP_011539851.1
XM_011541550.1 1684 Intron XP_011539852.1
XM_011541553.1 1684 Intron XP_011539855.1
XM_011541554.1 1684 Intron XP_011539856.1
XM_017001411.1 1684 Intron XP_016856900.1
XM_017001412.1 1684 Intron XP_016856901.1
XM_017001413.1 1684 Intron XP_016856902.1
XM_017001414.1 1684 Intron XP_016856903.1
XM_017001415.1 1684 Intron XP_016856904.1
XM_017001416.1 1684 Intron XP_016856905.1

View Full Product Details