Product Details

SNP ID
rs17849933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48077201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATAATCTCTACATCATCTTCAT[C/T]TTCTTTATTATGCCCAGGAGGTTCC
Phenotype
MIM: 605718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED4 PubMed Links

Gene Details

Gene
MED4
Gene Name
mediator complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270629.1 814 Missense Mutation AAT,GAT N205D NP_001257558.1
NM_014166.3 814 Missense Mutation AAT,GAT N251D NP_054885.1
XM_017020549.1 814 Missense Mutation AAT,GAT N229D XP_016876038.1
Gene
MED4-AS1
Gene Name
MED4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details