Product Details

SNP ID
rs17388251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123024708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCAGTTCCCAAACTTGTGTTAGA[C/T]AGAATCAGAAAAACAGGTAGTTGTG
Phenotype
MIM: 123842
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIC PubMed Links

Gene Details

Gene
PPIC
Gene Name
peptidylprolyl isomerase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000943.4 Intron NP_000934.1
Gene
SNX24
Gene Name
sorting nexin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014035.2 Intron NP_054754.1
XM_005271972.4 Intron XP_005272029.1
XM_006714592.3 Intron XP_006714655.1
XM_011543349.2 Intron XP_011541651.1
XM_011543350.2 Intron XP_011541652.1
XM_011543352.1 Intron XP_011541654.1
XM_017009394.1 Intron XP_016864883.1
XM_017009395.1 Intron XP_016864884.1
XM_017009396.1 Intron XP_016864885.1

View Full Product Details