Product Details

SNP ID
rs17148765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:122065369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAGATTTTAATACTGGTTAAT[A/G]TAGGAAAAATTATGCTTAAGCACCA
Phenotype
MIM: 153455 MIM: 610479
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOX PubMed Links

Gene Details

Gene
LOX
Gene Name
lysyl oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178102.2 1971 UTR 3 NP_001171573.1
NM_001317073.1 1971 UTR 3 NP_001304002.1
NM_002317.6 1971 UTR 3 NP_002308.2
Gene
SRFBP1
Gene Name
serum response factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152546.2 1971 Intron NP_689759.2
XM_017009111.1 1971 Intron XP_016864600.1

View Full Product Details