Product Details

SNP ID
rs16991171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34080635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCCTTTGGTTGCATGAATCTC[A/G]TCACGTCTCCCATCACATGGATCAC
Phenotype
MIM: 611973 MIM: 600444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS6 PubMed Links

Gene Details

Gene
MRPS6
Gene Name
mitochondrial ribosomal protein S6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032476.3 Intron NP_115865.1
Gene
SLC5A3
Gene Name
solute carrier family 5 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006933.6 Intron NP_008864.4

View Full Product Details