Product Details

SNP ID
rs16865448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177615804 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAAAGAATAAAGGAATTTCTGA[A/C]AGCCTAAAATTAGAAGGTATGGAAC
Phenotype
MIM: 604961
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PDE11A PubMed Links

Gene Details

Gene
PDE11A
Gene Name
phosphodiesterase 11A
There are no transcripts associated with this gene.

Gene
TTC30A
Gene Name
tetratricopeptide repeat domain 30A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152275.3 3163 UTR 3 NP_689488.3

View Full Product Details