Product Details

SNP ID
rs17744505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:229693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCTCAAGGCTCAGAAAAGCCGG[G/T]CTCGCGCGTGCTCTGCTGGCGGCCG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPL23AP53 PubMed Links

Gene Details

Gene
RPL23AP53
Gene Name
ribosomal protein L23a pseudogene 53
There are no transcripts associated with this gene.

Gene
ZNF596
Gene Name
zinc finger protein 596
There are no transcripts associated with this gene.

View Full Product Details