Product Details

SNP ID
rs7465247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:216997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACCTGACAAACAGTAGCTCTGC[A/C]AAATGATCCAAGTGAATATCAAAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927506 PubMed Links

Gene Details

Gene
LOC101927506
Gene Name
uncharacterized LOC101927506
There are no transcripts associated with this gene.

Gene
RPL23AP53
Gene Name
ribosomal protein L23a pseudogene 53
There are no transcripts associated with this gene.

View Full Product Details