Product Details
- SNP ID
-
rs17027754
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
32
- Location
-
Chr.1:111470803 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCTCCAGAGCTTGACAGGGTGTTT[G/T]CGTAGGTGCTAACTAAATGTCTGAT
- Phenotype
-
MIM: 603270
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP5F1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79859545] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP5F1
- Gene Name
- ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
There are no transcripts associated with this gene.
- Gene
- C1orf162
- Gene Name
- chromosome 1 open reading frame 162
View Full Product Details