Product Details

SNP ID
hCV33845966
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.19:41091747 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GCAACCCCAGCTCTATGAGATGTTC[C/T]CTTCGGTGATGAAACACCTGCCAGG
Phenotype
MIM: 608055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP2A13 PubMed Links

Gene Details

Gene
CYP2A13
Gene Name
cytochrome P450 family 2 subfamily A member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000766.4 Intron NP_000757.2

View Full Product Details