Product Details
- SNP ID
-
rs34163641
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:43614039 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTAGAACCTCTAGCAAGGAGAGC[A/G]ACCCCAGTCAATCTACAGTCCCCAG
- Phenotype
-
MIM: 609744
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CADM4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3815422] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CADM4
- Gene Name
- cell adhesion molecule 4
There are no transcripts associated with this gene.
- Gene
- SRRM5
- Gene Name
- serine/arginine repetitive matrix 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145641.1 |
1939 |
Missense Mutation |
AAC,GAC |
N640D |
NP_001139113.1 |
- Gene
- ZNF428
- Gene Name
- zinc finger protein 428
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182498.3 |
1939 |
Intron |
|
|
NP_872304.2 |
View Full Product Details