Product Details

SNP ID
rs17851581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22245215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTATTTGAGCATCTCTGAGCTG[C/T]AGCCTGAGGACGAGGCTATGTATTA
Phenotype
MIM: 605141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPREB1 PubMed Links

Gene Details

Gene
VPREB1
Gene Name
pre-B lymphocyte 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303509.1 336 Nonsense Mutation CAG,TAG Q105* NP_001290438.1
NM_007128.3 336 Nonsense Mutation CAG,TAG Q106* NP_009059.1

View Full Product Details