Product Details

SNP ID
rs17004894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24291272 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTACAGATTATGAAATGAAATTTT[C/G]GAACTTCTGAAAGCTACTAATCCAC
Phenotype
MIM: 614140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPECC1L PubMed Links

Gene Details

Gene
SPECC1L
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145468.3 Intron NP_001138940.3
NM_001254732.2 Intron NP_001241661.2
NM_001254733.1 Intron NP_001241662.1
NM_015330.4 Intron NP_056145.4
Gene
SPECC1L-ADORA2A
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details