Product Details

SNP ID
rs8136241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37837609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGAGTTTGGTATCTGAGTGGGGT[C/G]CTGGAACCAATCCCAAGAACACTGA
Phenotype
MIM: 613383
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD54 PubMed Links

Gene Details

Gene
ANKRD54
Gene Name
ankyrin repeat domain 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138797.2 Intron NP_620152.1
XM_006724136.1 Intron XP_006724199.1
XM_006724137.1 Intron XP_006724200.1
XM_011529877.2 Intron XP_011528179.1
XM_011529878.2 Intron XP_011528180.1
XM_011529879.2 Intron XP_011528181.1
XM_011529880.2 Intron XP_011528182.1
XM_011529882.2 Intron XP_011528184.1
XM_011529883.2 Intron XP_011528185.1
XM_011529884.2 Intron XP_011528186.1
XM_017028590.1 Intron XP_016884079.1
XM_017028591.1 Intron XP_016884080.1
XM_017028593.1 Intron XP_016884082.1
Gene
MIR658
Gene Name
microRNA 658
There are no transcripts associated with this gene.

View Full Product Details