Product Details

SNP ID
rs17115741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103597966 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGTCCTACACCTATTCTGCAAA[A/C]ACTAGTAATAGGAAAATAATGCCTG
Phenotype
MIM: 603804
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NEURL1 PubMed Links

Gene Details

Gene
NEURL1
Gene Name
neuralized E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
SH3PXD2A
Gene Name
SH3 and PX domains 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014631.2 10414 UTR 3 NP_055446.2
XM_005270294.4 10414 UTR 3 XP_005270351.1
XM_005270295.4 10414 UTR 3 XP_005270352.1
XM_005270297.3 10414 UTR 3 XP_005270354.1
XM_005270298.4 10414 UTR 3 XP_005270355.1
XM_006718079.3 10414 UTR 3 XP_006718142.1
XM_006718080.3 10414 UTR 3 XP_006718143.1
XM_011540385.2 10414 UTR 3 XP_011538687.1
XM_011540386.2 10414 UTR 3 XP_011538688.1
XM_011540387.2 10414 UTR 3 XP_011538689.1
XM_017016969.1 10414 UTR 3 XP_016872458.1
XM_017016970.1 10414 UTR 3 XP_016872459.1

View Full Product Details