Product Details

SNP ID
rs17121427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89431523 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATAGGCAAGCTCTCTTGCGGAGC[A/G]TACACATTAACTCAGGATCATCCTC
Phenotype
MIM: 611910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC16A12 PubMed Links

Gene Details

Gene
SLC16A12
Gene Name
solute carrier family 16 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213606.3 3281 UTR 3 NP_998771.3
XM_017016237.1 3281 UTR 3 XP_016871726.1
XM_017016238.1 3281 UTR 3 XP_016871727.1
XM_017016239.1 3281 UTR 3 XP_016871728.1

View Full Product Details