Product Details

SNP ID
rs17099029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:142594968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGTTACTAATGTCCCACTTAGC[C/T]GACCCCTTAACACACTTCATTTAGC
Phenotype
MIM: 131220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF1 PubMed Links

Gene Details

Gene
FGF1
Gene Name
fibroblast growth factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000800.4 717 UTR 3 NP_000791.1
NM_001144892.2 717 UTR 3 NP_001138364.1
NM_001144934.1 717 UTR 3 NP_001138406.1
NM_001144935.1 717 UTR 3 NP_001138407.1
NM_001257205.1 717 UTR 3 NP_001244134.1
NM_001257206.1 717 UTR 3 NP_001244135.1
NM_001257207.1 717 UTR 3 NP_001244136.1
NM_001257208.1 717 UTR 3 NP_001244137.1
NM_001257209.1 717 UTR 3 NP_001244138.1
NM_001257210.1 717 UTR 3 NP_001244139.1
NM_001257211.1 717 UTR 3 NP_001244140.1
NM_001257212.1 717 UTR 3 NP_001244141.1
NM_033136.3 717 UTR 3 NP_149127.1
NM_033137.2 717 UTR 3 NP_149128.1
XM_005268390.4 717 UTR 3 XP_005268447.1
XM_011537607.2 717 UTR 3 XP_011535909.1

View Full Product Details