Product Details
- SNP ID
-
rs7042533
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:88391011 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAATTATGCTCTTGCCAAACTACC[C/G]ACTGTTTTCTTTATATTCTTGCCCC
- Phenotype
-
MIM: 609936
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SPIN1
PubMed Links
Gene Details
- Gene
- SPIN1
- Gene Name
- spindlin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006717.2 |
|
Intron |
|
|
NP_006708.2 |
View Full Product Details