Product Details

SNP ID
rs16951818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50379068 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTACTGCCTCCTTAATCTACAC[A/G]TATTGCCAAAGTCTGCTTTTACTTA
Phenotype
MIM: 616673
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SKA1 PubMed Links

Gene Details

Gene
SKA1
Gene Name
spindle and kinetochore associated complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039535.2 Intron NP_001034624.1
NM_145060.3 Intron NP_659497.1

View Full Product Details