Product Details

SNP ID
rs17797932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:66503580 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCACACTTTCCACCAGTTTCATA[A/C]AGTTTGCATTTAATTATGAGTAAAT
Phenotype
MIM: 603016
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDH19 PubMed Links

Gene Details

Gene
CDH19
Gene Name
cadherin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271028.1 3737 UTR 3 NP_001257957.1
NM_021153.3 3737 UTR 3 NP_066976.1
XM_011525931.2 3737 UTR 3 XP_011524233.1
XM_011525932.1 3737 Intron XP_011524234.1
XM_017025711.1 3737 UTR 3 XP_016881200.1

View Full Product Details