Product Details

SNP ID
rs17032414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10116618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCTTCTTCTCCAAGTGTTTAA[C/G]AGAACTTGGCACTGGTTAGACTTTC
Phenotype
MIM: 611183
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRK1 PubMed Links

Gene Details

Gene
BRK1
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018462.4 Intron NP_060932.2
Gene
FANCD2OS
Gene Name
FANCD2 opposite strand
There are no transcripts associated with this gene.

View Full Product Details