Product Details
- SNP ID
-
rs17081968
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:70016696 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCTCCATCTGTCCTACATCTCAGA[G/T]AATGAAGTCTCTGCGAGGCTCACTG
- Phenotype
-
MIM: 610436
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RTTN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116199131] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RTTN
- Gene Name
- rotatin
View Full Product Details