Product Details

SNP ID
rs35002485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:121452484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGACCGGGGCTTCCTCGTGGT[G/T]CAGGGCGACCCGCGCGAGCTGCGGC
Phenotype
MIM: 602005
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105369536 PubMed Links

Gene Details

Gene
LOC105369536
Gene Name
uncharacterized LOC105369536
There are no transcripts associated with this gene.

Gene
SORL1
Gene Name
sortilin related receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003105.5 282 Silent Mutation GTG,GTT V51V NP_003096.1
XM_011542963.2 282 Silent Mutation GTG,GTT V51V XP_011541265.1
XM_011542965.2 282 Intron XP_011541267.1
XM_011542967.2 282 Intron XP_011541269.1
XM_017018169.1 282 Intron XP_016873658.1
XM_017018170.1 282 Intron XP_016873659.1
XM_017018171.1 282 Silent Mutation GTG,GTT V51V XP_016873660.1
XM_017018172.1 282 Intron XP_016873661.1

View Full Product Details