Product Details

SNP ID
rs35972846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:121452508 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGGGCGACCCGCGCGAGCTGCG[G/T]CTGTGGGCGCGCGGGGATGCCAGGG
Phenotype
MIM: 602005
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105369536 PubMed Links

Gene Details

Gene
LOC105369536
Gene Name
uncharacterized LOC105369536
There are no transcripts associated with this gene.

Gene
SORL1
Gene Name
sortilin related receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003105.5 306 Silent Mutation CGG,CGT R59R NP_003096.1
XM_011542963.2 306 Silent Mutation CGG,CGT R59R XP_011541265.1
XM_011542965.2 306 Intron XP_011541267.1
XM_011542967.2 306 Intron XP_011541269.1
XM_017018169.1 306 Intron XP_016873658.1
XM_017018170.1 306 Intron XP_016873659.1
XM_017018171.1 306 Silent Mutation CGG,CGT R59R XP_016873660.1
XM_017018172.1 306 Intron XP_016873661.1

View Full Product Details