Product Details

SNP ID
rs16982203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58656859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGCTCTGGGCTAAGCCCTTTCC[A/G]TGTGTAAGCTCAGCTCATCTTCACA
Phenotype
MIM: 603666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107985410 PubMed Links

Gene Details

Gene
LOC107985410
Gene Name
uncharacterized LOC107985410
There are no transcripts associated with this gene.

Gene
STX16
Gene Name
syntaxin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001433.2 Intron NP_001001433.1
NM_001134772.2 Intron NP_001128244.1
NM_001134773.2 Intron NP_001128245.1
NM_001204868.1 Intron NP_001191797.1
NM_003763.5 Intron NP_003754.2
Gene
STX16-NPEPL1
Gene Name
STX16-NPEPL1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details