Product Details
- SNP ID
-
rs16832418
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:121602317 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCACAAATCTGCATCTCCACATTC[C/T]AGTCCCTCATCTTCCCATCCATGAG
- Phenotype
-
MIM: 609107
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO40
PubMed Links
Gene Details
- Gene
- FBXO40
- Gene Name
- F-box protein 40
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016298.3 |
|
Intron |
|
|
NP_057382.2 |
View Full Product Details