Product Details
- SNP ID
-
rs17104100
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:67192047 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGACCGCAAGGGATCAGCTAGTGCT[A/G]GAACGAGCTCATCCATCTCAAATAA
- Phenotype
-
MIM: 603930
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM71D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60308909] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM71D
- Gene Name
- family with sequence similarity 71 member D
- Gene
- GPHN
- Gene Name
- gephyrin
View Full Product Details